Learn More
4-(4-Aminophenoxy)-2-pyridinecarboxamide, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(4-Aminophenoxy)-2-pyridinecarboxamide |
| CAS | 284462-80-2 |
| Molecular Formula | C12 H11 N3 O2 |
| Quantity | 50 mg |
| Synonym | 4-(2-Carbamoyl-4-pyridyloxy)aniline, 4-(4-Aminophenoxy)picolinamide |
| InChI Formula | InChI=1S/C12H11N3O2/c13-8-1-3-9(4-2-8)17-10-5-6-15-11(7-10)12(14)16/h1-7H,13H2,(H2,14,16) |
| SMILES | NC(=O)c1cc(Oc2ccc(N)cc2)ccn1 |
| IUPAC Name | 4-(4-aminophenoxy)pyridine-2-carboxamide |
| Molecular Weight (g/mol) | 229.235 |
| Formula Weight | 229.085 |