missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluorocrotonic acid, 97%, Thermo Scientific™

Catalog Number p-7077621
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
71027-02-6
C4H3F3O2
140.06
QZBAYURFHCTXOJ-OWOJBTEDSA-N
4,4,4-trifluorocrotonic acid, e-4,4,4-trifluorobut-2-enoic acid, 2e-4,4,4-trifluorobut-2-enoic acid, 4,4,4,-trifluorobut-2-enoic acid, 4,4,4-trifluoro-2-butenoic acid, hiddpndiaicichibejifaap, trifluorocrotonic acid, pubchem22644, 4,4,4-trifluoro crotonic acid
5709614
(E)-4,4,4-trifluorobut-2-enoic acid
C(=CC(F)(F)F)C(=O)O
This item is not returnable. View return policy
71027-02-6
C4H3F3O2
140.06
QZBAYURFHCTXOJ-OWOJBTEDSA-N
4,4,4-trifluorocrotonic acid, e-4,4,4-trifluorobut-2-enoic acid, 2e-4,4,4-trifluorobut-2-enoic acid, 4,4,4,-trifluorobut-2-enoic acid, 4,4,4-trifluoro-2-butenoic acid, hiddpndiaicichibejifaap, trifluorocrotonic acid, pubchem22644, 4,4,4-trifluoro crotonic acid
5709614
(E)-4,4,4-trifluorobut-2-enoic acid
C(=CC(F)(F)F)C(=O)O
Melting Point 53.0°C to 58.0°C
Boiling Point 68.0°C to 69.0°C (20.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 140.06
Percent Purity 97%
Chemical Name or Material 4,4,4-Trifluorocrotonic acid
compliance-icons
Product Identifier
  • 4,4,4-Trifluorocrotonic acid, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'