missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-2-butanone, 97%

Product Code. 11417074
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417074 1 g 1g
11427074 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417074 Supplier Thermo Scientific Alfa Aesar Supplier No. B21514.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2366-70-3 | C4H5F3O | 126.078 g/mol

It is used as a pharmaceutical intermediate. It is used for the synthesis of racemic 2-amino-5,5,5-trifluoro-3-methylpentanoic acid as fluorinated synthon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate. It is used for the synthesis of racemic 2-amino-5,5,5-trifluoro-3-methylpentanoic acid as fluorinated synthon.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges. Keep ignition sources away. Prevent formation of aerosols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2366-70-3
Molecular Formula C4H5F3O
Molecular Weight (g/mol) 126.078
MDL Number MFCD00077601
InChI Key BTXXTMOWISPQSJ-UHFFFAOYSA-N
Synonym 4,4,4-trifluoro-2-butanone, 2-butanone, 4,4,4-trifluoro, trifluoromethylacetone, trifluoromethyl acetone, acmc-1cb0f, 3-oxo-1,1,1-trifluorobutane
PubChem CID 539001
IUPAC Name 4,4,4-trifluorobutan-2-one
SMILES CC(=O)CC(F)(F)F

Specifications

Density 1.05
Boiling Point 95°C to 96°C
Flash Point 37°C (99°F)
Quantity 1 g
UN Number UN1224
Formula Weight 126.08
Percent Purity 97%
Chemical Name or Material 4,4,4-Trifluoro-2-butanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.