missing translation for 'onlineSavingsMsg'
Learn More

4,4',4″-Trichlorotrityl alcohol, 95%

Product Code. p-7044532
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11337076

Brand: Thermo Scientific Alfa Aesar L06965.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3010-80-8 | C19H13Cl3O | 363.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3010-80-8
Molecular Formula C19H13Cl3O
Molecular Weight (g/mol) 363.66
MDL Number MFCD00051795
InChI Key BPFKTJMHOWDJKI-UHFFFAOYSA-N
Synonym tris 4-chlorophenyl methanol, 4,4',4-trichlorotrityl alcohol, tris-4-chloro-phenyl-methanol, tri 4-chlorophenyl methanol, acmc-1clfy, tris-p-chlorophenyl methanol, tris 4-chlorophenyl methan-1-ol, 4,4,4-trichlorotrityl alcohol, benzenemethanol,4-chloro-a,a-bis 4-chlorophenyl
PubChem CID 76379
IUPAC Name tris(4-chlorophenyl)methanol
SMILES OC(C1=CC=C(Cl)C=C1)(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 94°C to 97°C
Quantity 2 g
Beilstein 2293156
Formula Weight 363.67
Percent Purity 95%
Chemical Name or Material 4,4',4''-Trichlorotrityl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.