Learn More
4-{[4-(4-Methyloxyphenyl)-piperazin-1-yl]-phenyl}-2,4-dihydro-[1,2,4]-triazol-3-one, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Dihydro-4-[4-[4-(4-methoxyphenyl)-1-piperazinyl]phenyl]-3H-1,2,4-triazol-3-one |
| CAS | 74853-07-9 |
| Molecular Formula | C19 H21 N5 O2 |
| Quantity | 100 mg |
| Synonym | 2,4-Dihydro-4-[4-[4-(4-methoxyphenyl)-1-piperazinyl]phenyl]-3H-1,2,4-triazol-3-one |
| InChI Formula | InChI=1S/C19H21N5O2/c1-26-18-8-6-16(7-9-18)23-12-10-22(11-13-23)15-2-4-17(5-3-15)24-14-20-21-19(24)25/h2-9,14H,10-13H2,1H3,(H,21,25) |
| SMILES | COc1ccc(cc1)N2CCN(CC2)c3ccc(cc3)N4C=NNC4=O |
| IUPAC Name | 4-[4-[4-(4-methoxyphenyl)piperazin-1-yl]phenyl]-1H-1,2,4-triazol-5-one |
| Molecular Weight (g/mol) | 351.4 |
| Formula Weight | 351.1695 |