missing translation for 'onlineSavingsMsg'
Learn More

4,4'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis-benzeneacetamide (Atenolol Impurity E), TRC

Product Code. 30010205
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010205 25 mg 25mg
30137951 100 mg 100mg
17924055 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010205 Supplier Toronto Research Chemicals Supplier No. TRCH94899525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141650-31-9
Formula Weight 358.1529
InChI Formula InChI=1S/C19H22N2O5/c20-18(23)9-13-1-5-16(6-2-13)25-11-15(22)12-26-17-7-3-14(4-8-17)10-19(21)24/h1-8,15,22H,9-12H2,(H2,20,23)(H2,21,24)
IUPAC Name 2-[4-[3-[4-(2-amino-2-oxoethyl)phenoxy]-2-hydroxypropoxy]phenyl]acetamide
Molecular Formula C19 H22 N2 O5
Molecular Weight (g/mol) 358.39
Percent Purity >95%
SMILES NC(=O)Cc1ccc(OCC(O)COc2ccc(CC(=O)N)cc2)cc1
Synonym 2,2'-[(2-Hydroxypropane-1,3-diyl)bis(oxy-4,1-phenylene)]diacetamide, Atenolol Imp. E (EP), Benzeneacetamide, 4,4'-[(2-hydroxy-1,3-propanediyl)bis(oxy)]bis-, 4,4'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis[benzeneacetamide]
Chemical Name or Material 2,2'-[(2-Hydroxypropane-1,3-diyl)bis(oxy-4,1-phenylene)]diacetamide
Grade HPLC
Show More Show Less

Specifications

Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.