Learn More
4-(3-Phenothiazin-10-ylpropyl)-1-piperazineethanol, TRC
Product Code. 30420086
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30420086 100 mg
30423424 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30420086 Supplier Toronto Research Chemicals Supplier No. TRCP295320100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3533-97-9
Molecular Formula C21 H27 N3 O S
Molecular Weight (g/mol) 369.52
Synonym 2-[4-[3-(10H-Phenothiazin-10-yl)propyl]piperazin-1-yl]ethanol, Perphenazine Imp. B (EP)
IUPAC Name 2-[4-(3-phenothiazin-10-ylpropyl)piperazin-1-yl]ethanol
SMILES OCCN1CCN(CCCN2c3ccccc3Sc4ccccc24)CC1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C21H27N3OS/c25-17-16-23-14-12-22(13-15-23)10-5-11-24-18-6-1-3-8-20(18)26-21-9-4-2-7-19(21)24/h1-4,6-9,25H,5,10-17H2
Formula Weight 369.1875
Percent Purity >95%
Chemical Name or Material 2-[4-[3-(10H-Phenothiazin-10-yl)propyl]piperazin-1-yl]ethanol
Grade HPLC