Learn More
4-(3-Oxopiperazin-1-yl)benzonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(3-Oxopiperazin-1-yl)benzonitrile |
| CAS | 1019601-14-9 |
| Molecular Formula | C11H11N3O |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C11H11N3O/c12-7-9-1-3-10(4-2-9)14-6-5-13-11(15)8-14/h1-4H,5-6,8H2,(H,13,15) |
| SMILES | O=C1CN(CCN1)c2ccc(cc2)C#N |
| IUPAC Name | 4-(3-oxopiperazin-1-yl)benzonitrile |
| Molecular Weight (g/mol) | 201.22 |
| Formula Weight | 201.0902 |