missing translation for 'onlineSavingsMsg'
Learn More

4-{[3-(chloromethyl)isoxazol-5-yl]methyl}morpholine, Tech., Thermo Scientific™

Product Code. 10587104
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10587104 1 g Amber Glass Bottle 1g
10683265 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10587104 Supplier Thermo Scientific Maybridge Supplier No. CC70824DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 953408-97-4
Molecular Formula C9H13ClN2O2
Molecular Weight (g/mol) 216.665
MDL Number MFCD11841078
InChI Key FZRKHZMWHFBKDV-UHFFFAOYSA-N
Synonym 4-3-chloromethyl isoxazol-5-yl methyl morpholine, 4-3-chloromethyl-1,2-oxazol-5-yl methyl morpholine, 3-chloromethyl-5-morpholin-4-yl methyl isoxazole
PubChem CID 43811054
IUPAC Name 4-[[3-(chloromethyl)-1,2-oxazol-5-yl]methyl]morpholine
SMILES C1COCCN1CC2=CC(=NO2)CCl

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 216.67
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-{[3-(Chloromethyl)isoxazol-5-yl]methyl}morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.