Learn More
4-(3-Aminopropyl)-N,N-dimethylaniline Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(3-Aminopropyl)-N,N-dimethylaniline Hydrochloride |
| CAS | 2741448-85-9 |
| Molecular Formula | C11 H18 N2 . H Cl |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C11H18N2.ClH/c1-13(2)11-7-5-10(6-8-11)4-3-9-12;/h5-8H,3-4,9,12H2,1-2H3;1H |
| SMILES | Cl.CN(C)c1ccc(CCCN)cc1 |
| IUPAC Name | 4-(3-aminopropyl)-N,N-dimethylaniline;hydrochloride |
| Molecular Weight (g/mol) | 214.735 |
| Formula Weight | 214.124 |