missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methylphenoxy)benzenesulfonyl chloride, 96%, Thermo Scientific™

Product Code. 15426387
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15426387

Brand: Thermo Scientific Alfa Aesar H33556.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610277-83-3
Molecular Formula C13H11ClO3S
Molecular Weight (g/mol) 282.74
MDL Number MFCD01631900
InChI Key IJPKGMOVNZHZKZ-UHFFFAOYSA-N
Synonym 4-2-methylphenoxy benzenesulfonyl chloride, 4-o-tolyloxyphenylsulfonyl chloride, 4-o-tolyloxy-benzenesulfonyl chloride, 4-o-tolyloxy benzenesulfonyl chloride, 4-o-tolyl-oxy-benzenesulfonyl chloride, 4-o-tolyloxy benzene-1-sulfonyl chloride, 4-2-methylphenoxy benzenesulphonyl chloride, 4-2-methylphenoxy benzene-1-sulfonyl chloride, 4-2-methylphenoxy phenylsulfonyl chloride, 4-2-methylphenoxy benzenesulfonylchloride
PubChem CID 2794712
IUPAC Name 4-(2-methylphenoxy)benzenesulfonyl chloride
SMILES CC1=CC=CC=C1OC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 81°C to 85°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 282.74
Percent Purity 96%
Chemical Name or Material 4-(2-Methylphenoxy)benzenesulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.