Learn More
4-(2-Ethoxy-2-oxoethoxy)phenylboronic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(2-Ethoxy-2-oxoethoxy)phenylboronic Acid |
| CAS | 282116-97-6 |
| Molecular Formula | C10H13BO5 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C10H13BO5/c1-2-15-10(12)7-16-9-5-3-8(4-6-9)11(13)14/h3-6,13-14H,2,7H2,1H3 |
| SMILES | CCOC(=O)COc1ccc(cc1)B(O)O |
| IUPAC Name | [4-(2-ethoxy-2-oxoethoxy)phenyl]boronic acid |
| Molecular Weight (g/mol) | 224.02 |
| Formula Weight | 224.0856 |