missing translation for 'onlineSavingsMsg'
Learn More

4-[(2-Chloro-6-fluorobenzyl)oxy]-3-methoxybenzaldehyde, 97%, Thermo Scientific™

Product Code. 10572632
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10572632 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10572632 Supplier Thermo Scientific Maybridge Supplier No. AW01128DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 306934-75-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C15H12ClFO3
MDL Number MFCD01590672
Quantity 1 g
Synonym 4-2-chloro-6-fluorobenzyl oxy-3-methoxybenzaldehyde, 4-2-chloro-6-fluorophenyl methoxy-3-methoxybenzaldehyde, 4-2-chloro-6-fluorobenzyloxy-3-methoxybenzaldehyde, benzaldehyde,4-2-chloro-6-fluorophenyl methoxy-3-methoxy, pubchem2893, cambridge id 5900464, 2-2-chloro-6-fluorobenzyl oxy-5-formylanisole, 4-6-chloro-2-fluorophenyl methoxy-3-methoxybenzaldehyde, 4-2-chloranyl-6-fluoranyl-phenyl methoxy-3-methoxy-benzaldehyde
InChI Key GYBXSWQBJQILED-UHFFFAOYSA-N
SMILES COC1=C(C=CC(=C1)C=O)OCC2=C(C=CC=C2Cl)F
IUPAC Name 4-[(2-chloro-6-fluorophenyl)methoxy]-3-methoxybenzaldehyde
Molecular Weight (g/mol) 294.706
PubChem CID 735741
Formula Weight 294.71
Percent Purity 97%
Chemical Name or Material 4-[(2-Chloro-6-fluorobenzyl)oxy]-3-methoxybenzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.