Learn More
4-(2-Carboxyethyl)benzoic Acid Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(2-Carboxyethyl)benzoic Acid Methyl Ester |
| CAS | 151937-09-6 |
| Molecular Formula | C11H12O4 |
| Quantity | 1 g |
| Synonym | Methyl 4-(2-Carboxyethyl)benzoate, 3-[4-(Methoxycarbonyl)phenyl]propanoic Acid |
| InChI Formula | InChI=1S/C11H12O4/c1-15-11(14)9-5-2-8(3-6-9)4-7-10(12)13/h2-3,5-6H,4,7H2,1H3,(H,12,13) |
| SMILES | COC(=O)c1ccc(CCC(=O)O)cc1 |
| IUPAC Name | 3-(4-methoxycarbonylphenyl)propanoic acid |
| Molecular Weight (g/mol) | 208.21 |
| Formula Weight | 208.0736 |