Learn More
4-(2,4-Dichloro-5-methoxyphenoxy)aniline, TRC
Product Code. 30381716
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30381716 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30381716 Supplier Toronto Research Chemicals Supplier No. TRCD43507525MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(2,4-Dichloro-5-methoxyphenoxy)aniline
CAS 125138-50-3
Molecular Formula C13H11Cl2NO2
Quantity 25 mg
Synonym 4-(2,4-Dichloro-5-methoxyphenoxy)benzenamine
InChI Formula InChI=1S/C13H11Cl2NO2/c1-17-12-7-13(11(15)6-10(12)14)18-9-4-2-8(16)3-5-9/h2-7H,16H2,1H3
SMILES ClC1=C(OC2=CC=C(N)C=C2)C=C(OC)C(Cl)=C1
IUPAC Name 4-(2,4-dichloro-5-methoxyphenoxy)aniline
Molecular Weight (g/mol) 284.14
Formula Weight 283.02