Learn More
4-(2-(4-Chlorobenz-d4-amido)ethyl)phenyl 4-Chlorobenz-d4-oate, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(2-(4-Chlorobenz-d4-amido)ethyl)phenyl 4-Chlorobenz-d4-oate |
| CAS | 2733570-40-4 |
| Molecular Formula | C22 D8 H9 Cl2 N O3 |
| Quantity | 25 mg |
| Synonym | 4-[2-[(4-Chlorobenz-d4-oyl)amino]ethyl]phenyl Ester 4-Chlorobenzoic-d4 Acid |
| InChI Formula | InChI=1S/C22H17Cl2NO3/c23-18-7-3-16(4-8-18)21(26)25-14-13-15-1-11-20(12-2-15)28-22(27)17-5-9-19(24)10-6-17/h1-12H,13-14H2,(H,25,26)/i3D,4D,5D,6D,7D,8D,9D,10D |
| SMILES | [2H]c1c([2H])c(C(=O)NCCc2ccc(OC(=O)c3c([2H])c([2H])c(Cl)c([2H])c3[2H])cc2)c([2H])c([2H])c1Cl |
| IUPAC Name | [4-[2-[(4-chloro-2,3,5,6-tetradeuteriobenzoyl)amino]ethyl]phenyl] 4-chloro-2,3,5,6-tetradeuteriobenzoate |
| Molecular Weight (g/mol) | 422.331 |
| Formula Weight | 421.109 |