missing translation for 'onlineSavingsMsg'
Learn More

4'-(1-Imidazolyl)acetophenone, 97%, Thermo Scientific™

Product Code. p-7044385
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352386 5 g 5g
11362386 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352386 Supplier Thermo Scientific Alfa Aesar Supplier No. L06115.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10041-06-2
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD00005282
InChI Key GAIQQJIMVVUTQN-UHFFFAOYSA-N
Synonym 4-imidazol-1-yl acetophenone, 4'-imidazol-1-yl acetophenone, 1-4-1h-imidazol-1-yl phenyl ethanone, 4'-1-imidazolyl acetophenone, 1-4-imidazol-1-yl-phenyl-ethanone, 1-4-acetylphenyl imidazole, ethanone, 1-4-1h-imidazol-1-yl phenyl, 1-4-imidazol-1-ylphenyl ethanone, 1-4-1h-imidazol-1-yl phenyl ethan-1-one, 4'-1h-imidazol-1-yl acetophenone
PubChem CID 82316
IUPAC Name 1-(4-imidazol-1-ylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)N2C=CN=C2

Specifications

Melting Point 114°C to 118°C
Quantity 5 g
Beilstein 608955
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 4'-(1-Imidazolyl)acetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.