missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3,3-Trimethyl-7-oxabicyclo[4.1.0]hept-2-yl)-3-buten-2-one, tech., Thermo Scientific™

Product Code. 15420656
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420656 5 g 5g
15410656 1 g 1g
15430656 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15420656 Supplier Thermo Scientific Alfa Aesar Supplier No. H53499.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-(1,3,3-Trimethyl-7-oxabicyclo[4.1.0]hept-2-yl)-3-buten-2-one is used as a pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 190059-33-7
Molecular Formula C13H20O2
Molecular Weight (g/mol) 208.30
MDL Number MFCD07784337
InChI Key ODMUHAHUBCUABS-UHFFFAOYNA-N
Synonym 4-1,3,3-trimethyl-7-oxabicyclo 4.1.0 hept-2-yl-3-buten-2-one, 4-1,3,3-trimethyl-7-oxabicyclo 4.1.0 heptan-2-yl but-3-en-2-one, 3-buten-2-one,4-1,3,3-trimethyl-7-oxabicyclo 4.1.0 hept-2-yl-, 3e, 4-1,3,3-trimethyl-7-oxabicyclo 4.1.0 hept-2-yl-3-buten-2-one, mixture of cis and trans
PubChem CID 11041982
SMILES CC(=O)C=CC1C2(C)OC2CCC1(C)C

Specifications

Density 1.039
Refractive Index 1.491
Quantity 5 g
IUPAC Name 4-{1,3,3-trimethyl-7-oxabicyclo[4.1.0]heptan-2-yl}but-3-en-2-one
Formula Weight 208.3
Grade Technical
Chemical Name or Material 4-(1,3,3-Trimethyl-7-oxabicyclo[4.1.0]hept-2-yl)-3-buten-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.