Learn More
(3S)-1,2,3,4-Tetrahydro-6,7-dimethoxy-3-isoquinolinecarboxylic Acid Hydrochloride, TRC
Product Code. 30423509
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423509 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30423509 Supplier Toronto Research Chemicals Supplier No. TRCT293190100MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (3S)-1,2,3,4-Tetrahydro-6,7-dimethoxy-3-isoquinolinecarboxylic acid hydrochloride
CAS 82586-62-7
Molecular Formula C12 H15 N O4 . Cl H
Quantity 100 mg
InChI Formula InChI=1S/C12H15NO4.ClH/c1-16-10-4-7-3-9(12(14)15)13-6-8(7)5-11(10)17-2;/h4-5,9,13H,3,6H2,1-2H3,(H,14,15);1H/t9-;/m0./s1
SMILES Cl.COc1cc2CN[C@@H](Cc2cc1OC)C(=O)O
IUPAC Name (3S)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid;hydrochloride
Molecular Weight (g/mol) 273.713
Formula Weight 273.077