missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Thunder-Link (R) PLUS oligo Antibody Conjugation Kit

Product Code. 18760804 Shop All Bio Techne Products
Click to view available options
Quantity:
300 μg
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18760804

Brand: Novus Biologicals™ 4250300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For simple and rapid conjugation of antibodies to oligonucleotides

Warm the Thunder-Link PLUS Conjugate Clean Up Reagent by placing the tube in warm water (not warmer than 40°C) for 10 minutes, then shaking to re-dissolve the contents. Once dissolved, maintain the tube at approx.22° C to prevent any crystal formation before use.
TRUSTED_SUSTAINABILITY

Specifications

Description Antibody Labeling Kit
Content And Storage Store at 4°C. Do not freeze.
Quantity 300 μg
Target Thunder-Link PLUS oligo
For Use With (Application) Warm the Thunder-Link PLUS Conjugate Clean Up Reagent by placing the tube in warm water (not warmer than 40C) for 10 minutes, then shaking to re-dissolve the contents. Once dissolved, maintain the tube at ∼22C to prevent any crystal formation before use.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.