Learn More
(3R,6R)-3,6-Diphenyl-2,5-piperazinedione, TRC
Specifications
Specifications
| Chemical Name or Material | (3R,6R)-3,6-Diphenyl-2,5-piperazinedione |
| CAS | 31485-02-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C16 H14 N2 O2 |
| Quantity | 50 mg |
| Synonym | (3R,6R)-3,6-Diphenyl-2,5-piperazinedione, 2,5-Piperazinedione, 3,6-diphenyl-, D,D- (8CI), (3R,6R)-3,6-Diphenylpiperazine-2,5-dione |
| InChI Formula | InChI=1S/C16H14N2O2/c19-15-13(11-7-3-1-4-8-11)17-16(20)14(18-15)12-9-5-2-6-10-12/h1-10,13-14H,(H,17,20)(H,18,19)/t13-,14-/m1/s1 |
| SMILES | O=C1N[C@@H](C(=O)N[C@@H]1c2ccccc2)c3ccccc3 |
| IUPAC Name | (3R,6R)-3,6-diphenylpiperazine-2,5-dione |
| Molecular Weight (g/mol) | 266.29 |
| Show More |