Learn More
3PO, MedChemExpress
Product Code. 16450398
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16450398
Brand: Medchem Express HY19824
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress 3PO is a inhibitor of (PFKFB3). 3PO attenuates the proliferation of several cancer cell lines with IC50s of 1.4-24 μmol/L. 3PO suppresses glucose uptake and decreases the intracellular concentra tion of Fru-2,6-BP, lactate, ATP, NAD+, and NADH. 3PO can be used for the research of cancer.
3PO is a inhibitor of (PFKFB3). 3PO attenuates the proliferation of several cancer cell lines with IC50s of 1.4-24 μmol/L. 3PO suppresses glucose uptake and decreases the intracellular concentra tion of Fru-2,6-BP, lactate, ATP, NAD+, and NADH. 3PO can be used for the research of cancer.
Specifications
Chemical Name or Material 3PO
Physical Form Solid
CAS 18550-98-6
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C13H10N2O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 60 mg/mL (285.40 mM; Need ultrasonic)
SMILES O=C(C1=CC=NC=C1)/C=C/C2=CC=CN=C2
Molecular Weight (g/mol) 210.23
Formula Weight 210.23
Percent Purity 98.53%
Grade Research