Learn More
3Beta,5Alpha,6Beta-Trihydroxycholestane-d7, TRC
Specifications
Specifications
| Chemical Name or Material | 3beta,5alpha,6beta-Trihydroxycholestane-d7 |
| CAS | 127684-07-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C27H41D7O3 |
| Quantity | 1 mg |
| Synonym | (3β,5α,6β)-Cholestane-3,5,6-triol-d7, 3β-Hydroxycholestane-5α,6β-diol-d7, 5α,6β-Dihydroxycholestanol-d7, Cholesta-3β,5α,6β-triol-d7, Cholestane-3β,5α,6β-triol-d7, NSC 124751-d7, NSC 18178-d7 |
| InChI Formula | InChI=1S/C27H48O3/c1-17(2)7-6-8-18(3)21-9-10-22-20-15-24(29)27(30)16-19(28)11-14-26(27,5)23(20)12-13-25(21,22)4/h17-24,28-30H,6-16H2,1-5H3/t18-,19+,20+,21-,22+,23+,24-,25-,26-,27+/m1/s1/i1D3,2D3,17D |
| SMILES | O[C@H]1CC[C@@]2(C)[C@]([C@H](O)C[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CCCC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])(O)C1 |
| Molecular Weight (g/mol) | 427.71 |
| Formula Weight | 427.4043 |
| Show More |