Learn More
(3Beta)-3-O-tert-Butyldimethylsilyl-cholest-5-ene-3,24-diol, TRC
Specifications
Specifications
| Chemical Name or Material | (3beta)-3-O-tert-Butyldimethylsilyl-cholest-5-ene-3,24-diol |
| CAS | 84529-86-2 |
| Molecular Formula | C30H54O2Si |
| Quantity | 10 mg |
| Synonym | (3β)-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-chol-5-en-24-ol |
| InChI Formula | InChI=1S/C30H54O2Si/c1-21(10-9-19-31)25-13-14-26-24-12-11-22-20-23(32-33(7,8)28(2,3)4)15-17-29(22,5)27(24)16-18-30(25,26)6/h11,21,23-27,31H,9-10,12-20H2,1-8H3/t21?,23-,24-,25+,26-,27-,29?,30?/m0/s1 |
| SMILES | [H][C@@]12CC=C3C[C@@H](O[Si](C)(C)C(C)(C)C)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4([H])[C@H](C)CCCO |
| IUPAC Name | (R)-4-((3S,8S,9S,10R,13R,14S,17R)-3-((tert-butyldimethylsilyl)oxy)-10,13-dimethyl-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentan-1-ol |
| Molecular Weight (g/mol) | 474.83 |
| Formula Weight | 474.3893 |