Learn More
Greiner Bio-One 384-well Storage Microplates
GREENER_CHOICE
Product Code. 13018023
Click to view available options
Color:
Clear
Unit Size:
Pack of 80
Product Code. 13018023
Brand: Greiner Bio-One 788860906
Only null left
Add to basket
Add to basket
Reduce volume of liquid necessary for acoustic liquid handling with minimal dead volume.
  • Made from cycloolefin
  • DMSO resistant for compound storage
  • Small volume LoBase
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Description DMSO resistant
Array 16 x 24
Color Clear
Material Cycloolefin Copolymer
No. of Wells 384
Sterility Non-sterile
Well Shape Flat Bottom (F-well)