Learn More
Greiner Bio-One 384-well Storage Microplates
Click to view available options
Color:
Clear
Unit Size:
Pack of 80
Description
- Made from cycloolefin
- DMSO resistant for compound storage
- Small volume LoBase
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Description | DMSO resistant |
| Array | 16 x 24 |
| Color | Clear |
| Material | Cycloolefin Copolymer |
| No. of Wells | 384 |
| Sterility | Non-sterile |
| Well Shape | Flat Bottom (F-well) |