Learn More
Corning™ 384-Well Black/Clear Round Bottom Ultra-Low Attachment Spheroid Microplate with Generic Bar codes, With Lid, Sterile
Alle ansehen Corning Produkte
Beschreibung
- Surface minimizes protein absorption, enzyme activation and cellular activation
- Surface is noncytotoxic, biologically inert and nondegradable
- Sterilized by gamma irradiation
- Black wall microplates have low background fluorescence, minimal light scatter and reduced crosstalk
- Opaque walls to prevent well-to-well crosstalk
- Can be used for both top and bottom reading instruments
- Bulk packed 10 per bag
- Novel well geometry aids formation of spheroids in center of well
- Optically clear round bottom with black opaque microplate body
Spezifikation
Spezifikation
| Barcode | Includes Barcode |
| Lid | With Lid |
| Color | Black, Clear Bottom |
| Volume (Metric) Well | 90 μL |
| No. of Wells | 384 |
| Volume (Metric) Working | 20 to 80 μL |
| Sterility | Sterile |
| Surface Treatment | Ultra-low Binding |
| Well Shape | Round |
| Quantity | 10/Pk., 50/Cs. |