Learn More
Applied Biosystems™ Sequencing Standard, BigDye™ Terminator v1.1 Dx
Shop All Applied Biosystems ProductsDescription
Includes
This kit contains four tubes of DNA of a known 1200 base pair sequence prepared with BigDye® Terminator v1.1. Each tube can perform one spectral calibration/performance check and/or one control sequencing run on the Applied Biosystems™ Dx Series Genetic Analyzers
Sequencing Standard, BigDye Terminator v1.1 Dx, consists of four tubes, each of which contains DNA of a known 1,200 base pair sequence prepared with BigDye Terminator v1.1. Sufficient DNA is included to perform one spectral calibration/performance check and/or one control sequencing run on an Applied Biosystems Dx genetic analyzer. The DNA preparation has been lyophilized to maximize stability.
Note: Applied Biosystems Dx genetic analyzers and consumables are for distribution and use in specific European countries only. Not for use in the USA.
Order Info
Each
Specifications
Specifications
| Content And Storage | This kit contains four tubes of DNA of a known 1200 base pair sequence prepared with BigDye™ Terminator v1.1. Each tube can perform one spectral calibration/performance check and/or one control sequencing run on the Applied Biosystems 3500 Dx Series Genetic Analyzers. Store kit at -15 to -25°C. |
| For Use With (Equipment) | 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer |
| For Use With (Application) | Sequencing |
| Label or Dye | ROX, TAMRA™, Rhodamine 110, Rhodamine 6G |
| Product Line | BigDye™ Terminator |
| Product Type | Sequencing Standard |
| Quantity | 4 Tubes |
For In Vitro Diagnostic Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.