Learn More
4-Amino-2-chlorobenzoic acid, 98%, BLD Pharm™
Product Code. 30948108
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
Unit Size:
100g
1g
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948108 5 g 5g
30935441 1 g 1g
30941999 100 g 100g
30942359 500 g 500g
30947378 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30948108 Supplier BLD Pharm Supplier No. BD104405g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 2457-76-3
Molecular Formula C7H6ClNO2
MDL Number MFCD00007772
Quantity 5 g
SMILES C1=CC(=C(C=C1N)Cl)C(O)=O
Molecular Weight (g/mol) 171.58
Percent Purity 98%
Chemical Name or Material 4-Amino-2-chlorobenzoic acid