missing translation for 'onlineSavingsMsg'
Learn More

PCIF1 Antibody [DyLight 405], Novus Biologicals Biologicals™

Product Code. 30500440 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30500440 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30500440 Supplier Novus Biologicals Supplier No. NBP337957V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PCIF1 Polyclonal antibody specifically detects PCIF1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCIF1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias bA465L10.1, C20orf67, chromosome 20 open reading frame 67, PDX-1 C terminus-interacting factor 1, PDX1 C-terminal inhibiting factor 1, phosphorylated CTD-interacting factor 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human PCIF1 (NP_071387.1).,, Sequence:, MANENHGSPREEASLLSHSPGTSNQSQPCSPKPIRLVQDLPEELVHAGWEKCWSRRENRPYYFNRFTNQSLWEMPVLGQHDVISDPLGLNATPLPQDSSLVETPPAENKPRKRQLSEEQPSGNGVKKPKIEIPVTPTGQSVPSSPSIPGTPTLKMWGTSPEDKQQAALLRPTEVYWDLDIQTNAVIKHRGPSEVLPPHPE
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63935
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.