missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LARP4 Antibody [Alexa Fluor« 532], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38122AF532
This item is not returnable.
View return policy
Description
LARP4 Polyclonal antibody specifically detects LARP4 in Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| LARP4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 113251 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Alexa Fluor 532 | |
| c-Mpl binding protein, DKFZp686E039, La ribonucleoprotein domain family member 4, La ribonucleoprotein domain family, member 4, la-related protein 4, MGC74631, PP13296 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human LARP4 (NP_443111.4).,, Sequence:, MLLFVEQVASKGTGLNPNAKVWQEIAPGNTDATPVTHGTESSWHEIAATSGAHPEGNAELSEDICKEYEVMYSSSCETTRNTTGIEESTDGMILGPEDLSYQIYDVSGESNSAVSTEDLK | |
| 0.1 mL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction