missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karyopherin (importin) beta 3 Antibody [DyLight 405], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35908V
This item is not returnable.
View return policy
Description
Karyopherin (importin) beta 3 Polyclonal antibody specifically detects Karyopherin (importin) beta 3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Karyopherin (importin) beta 3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3843 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot | |
| DyLight 405 | |
| DKFZp686O1576, FLJ43041, IMB3, imp5, importin 5, importin beta-3 subunit, Importin subunit beta-3, importin-5, karyopherin (importin) beta 3, Karyopherin beta-3, KPNB3, MGC2068, RAN binding protein 5, Ran_GTP binding protein 5, Ran-binding protein 5, RANBP5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-300 of human Karyopherin (importin) beta 3 (NP_002262.3).,, Sequence:, AAAAAEQQQFYLLLGNLLSPDNVVRKQAEETYENIPGQSKITFLLQAIRNTTAAEEARQMAAVLLRRLLSSAFDEVYPALPSDVQTAIKSELLMIIQMETQSSMRKKVCDIAAELARNLIDEDGNNQWPEGLKFLFDSVSSQNVGLREAALHIFWNFPGIFGNQQQHYLDVIKRMLVQCMQDQEHPSIRTLSARATAAFILANEHNVALFKHFADLLPGFLQAVNDSCYQNDDSVLKSLVEIADTVPKYLRPHLEATLQLSLKLCGDTSLNNMQRQLALEV | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction