missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MLC1SA Polyclonal antibody specifically detects MLC1SA in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | MLC1SA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | MLC1sa, MLC1SAmyosin light chain 1, slow-twitch muscle A isoform, myosin alkali light chain 1 slow a, myosin light chain 1 slow a, Myosin light chain 1 slow-twitch muscle A isoform, myosin light chain 6B, myosin, light chain 6B, alkali, smooth muscle and non-muscle, myosin, light polypeptide 6B, alkali, smooth muscle and non-muscle, smooth muscle and non-muscle myosin alkali light chain 6B, Smooth muscle and nonmuscle myosin light chain alkali 6B |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-208 of human MLC1SA (NP_002466.1).,, Sequence:, MPPKKDVPVKKPAGPSISKPAAKPAAAGAPPAKTKAEPAVPQAPQKTQEPPVDLSKVVIEFNKDQLEEFKEAFELFDRVGDGKILYSQCGDVMRALGQNPTNAEVLKVLGNPKSDELKSRRVDFETFLPMLQAVAKNRGQGTYEDYLEGFRVFDKEGNGKVMGAELRHVLTTLGEKMTEEEVETVLAGHEDSNGCINYEAFLKHILSV |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?