missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF331 Polyclonal antibody specifically detects ZNF331 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | ZNF331 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ZFN533, zinc finger protein 385B, Zinc finger protein 533FLJ25270, ZNF533 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 50-130 of human ZNF331 (NP_061025.5).,, Sequence:, AYENKSLPTEKNIHEIRASKRNSDRRSKSLGRNWICEGTLERPQRSRGRYVNQMIINYVKRPATREGTPPRTHQRHHKENS |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?