missing translation for 'onlineSavingsMsg'
Learn More

POLR1C Antibody [CoraFluorÖ 1], Novus Biologicals Biologicals™

Product Code. 30498106 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498106 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498106 Supplier Novus Biologicals Supplier No. NBP335071CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

POLR1C Polyclonal antibody specifically detects POLR1C in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR1C
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias AC40, DNA-directed RNA polymerase I subunit C, DNA-directed RNA polymerases I and III 40 kDa polypeptide, POLR1E, polymerase (RNA) I polypeptide C, 30kDa, RNA polymerase I subunit, RPA39RNA polymerases I and III subunit AC1, RPA40TCS3, RPA5RPAC1DNA-directed RNA polymerases I and III subunit RPAC1, RPC40
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human POLR1C (NP_976035.1).,, Sequence:, MAASQAVEEMRSRVVLGEFGVRNVHTTDFPGNYSGYDDAWDQDRFEKNFRVDVVHMDENSLEFDMVGIDAAIANAFRRILLAEVPTMAVEKVLVYNNTSIVQDEILAHRLGLIPIHADPRLFEYRNQGDEEGTEIDTLQFRLQVRCTRNPHAAKDSSDPNELYVNHKVYTRHMTWIPLGNQADLFPEGTIRPVHDDILIA
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 9533
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.