Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Custom Needles with Point 5 or AS
Product Code. p-8007080
Click to view available options
Needle Gauge:
17 G
19 G
Unit Size:
Pack of 6
Product Code. 11583254
Brand: Hamilton™ 775108
Please to purchase this item. Need a web account? Register with us today!
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
  • Available in various gauges
  • Autoclavable
  • Metal hubs manufactured from nickel plated brass
  • Custom length
  • Compatible with PTFE Luer Lock (TLL) syringes
  • Design controlled by a series of universal standards which guarantees compatibility between manufacturers
  • Provided in packs of 6
Specifications
Material Nickel-plated Brass (Hub), Stainless Steel (304)
Needle Gauge 17 G
Diameter (Metric) Inner 1.07 mm
Diameter (Metric) Outer 1.47 mm
Needle Point Style 5 or AS
For Use With (Equipment) TLL Syringes and LT or TLL Connectors