Learn More
Kinesis™ 3 way connector integral valves single T PTFE body 50psi 0.8mm bore size Omnifit
Brand: Kinesis™ 002422
Additional Details : Weight : 0.10000kg
Brand: Kinesis™ 002422
Additional Details : Weight : 0.10000kg