missing translation for 'onlineSavingsMsg'
Learn More

3-Undecyne, 97%, Thermo Scientific™

Product Code. 11490367
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11490367 5 g 5g
11400377 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11490367

Brand: Thermo Scientific Alfa Aesar B21753.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60212-30-8
Molecular Formula C11H20
Molecular Weight (g/mol) 152.281
MDL Number MFCD00041663
InChI Key DPWGJNPCPLQVKQ-UHFFFAOYSA-N
Synonym 3-undecyne, acmc-209mii, dpwgjnpcplqvkq-uhfffaoysa
PubChem CID 143689
IUPAC Name undec-3-yne
SMILES CCCCCCCC#CCC

Specifications

Density 0.77
Boiling Point 79°C to 81°C (10 mmHg)
Quantity 5 g
Formula Weight 152.28
Percent Purity 97%
Chemical Name or Material 3-Undecyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.