Learn More
3-(Trimethylsilyl)-1-propane-1,1,2,2,3,3-d6-sulfonic Acid Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(Trimethylsilyl)-1-propane-1,1,2,2,3,3-d6-sulfonic Acid Sodium Salt |
| CAS | 284664-85-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6 2H6 H9 O3 S Si . Na |
| Quantity | 50 mg |
| Synonym | 4,4-Dimethyl-4-(Silapentane-D6)-1-Sulfonic Acid |
| InChI Formula | InChI=1S/C6H16O3SSi.Na/c1-11(2,3)6-4-5-10(7,8)9;/h4-6H2,1-3H3,(H,7,8,9);/q;+1/p-1/i4D2,5D2,6D2; |
| SMILES | [Na+].[2H]C([2H])(C([2H])([2H])[Si](C)(C)C)C([2H])([2H])S(=O)(=O)[O-] |
| IUPAC Name | sodium;1,1,2,2,3,3-hexadeuterio-3-trimethylsilylpropane-1-sulfonate |
| Molecular Weight (g/mol) | 224.36 |
| Show More |