Learn More
3-Trifluoromethylphenylhydrazine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Trifluoromethylphenylhydrazine Hydrochloride |
| CAS | 3107-33-3 |
| Molecular Formula | C7H8ClF3N2 |
| Quantity | 10 g |
| Synonym | (alpha,alpha,alpha-trifluoro-m-tolyl)hydrazine Hydrochloride, [3-(trifluoromethyl)phenyl]hydrazine Monohydrochloride |
| InChI Formula | InChI=1S/C7H7F3N2.ClH/c8-7(9,10)5-2-1-3-6(4-5)12-11;/h1-4,12H,11H2;1H |
| SMILES | Cl.NNc1cccc(c1)C(F)(F)F |
| IUPAC Name | [3-(trifluoromethyl)phenyl]hydrazine;hydrochloride |
| Molecular Weight (g/mol) | 212.6 |
| Formula Weight | 212.0328 |