Learn More
3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride, 96%, Thermo Scientific™
Product Code. 15468845
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468845 250 mg 250mg
15458845 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15468845 Supplier Thermo Scientific Alfa Aesar Supplier No. H34235.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 885267-96-9
Molecular Formula C13H8ClF3O2S
Molecular Weight (g/mol) 320.71
MDL Number MFCD06409268
InChI Key WJSSWLPZDVPLIK-UHFFFAOYSA-N
Synonym 3'-trifluoromethyl biphenyl-4-sulfonyl chloride, 4-3-trifluoromethyl phenyl benzenesulfonyl chloride, 3'-trifluoromethyl-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-trifluoromethyl, 3'-trifluoromethyl 1,1'-biphenyl-4-sulfonyl chloride, 3'-trifluoromethylbiphenyl-4-sulfonyl chloride
PubChem CID 16244690
SMILES FC(F)(F)C1=CC=CC(=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
Specifications
Melting Point 78°C to 92°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
IUPAC Name 4-[3-(trifluoromethyl)phenyl]benzenesulfonyl chloride
Formula Weight 320.71
Percent Purity 96%
Chemical Name or Material 3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride