missing translation for 'onlineSavingsMsg'
Learn More

3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride, 96%, Thermo Scientific™

Product Code. 15458845
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458845 1 g 1g
15468845 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458845 Supplier Thermo Scientific Alfa Aesar Supplier No. H34235.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885267-96-9
Molecular Formula C13H8ClF3O2S
Molecular Weight (g/mol) 320.71
MDL Number MFCD06409268
InChI Key WJSSWLPZDVPLIK-UHFFFAOYSA-N
Synonym 3'-trifluoromethyl biphenyl-4-sulfonyl chloride, 4-3-trifluoromethyl phenyl benzenesulfonyl chloride, 3'-trifluoromethyl-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-trifluoromethyl, 3'-trifluoromethyl 1,1'-biphenyl-4-sulfonyl chloride, 3'-trifluoromethylbiphenyl-4-sulfonyl chloride
PubChem CID 16244690
SMILES FC(F)(F)C1=CC=CC(=C1)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 78°C to 92°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
IUPAC Name 3'-(trifluoromethyl)-[1,1'-biphenyl]-4-sulfonyl chloride
Formula Weight 320.71
Percent Purity 96%
Chemical Name or Material 3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.