missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)anisole, 98%, Thermo Scientific Chemicals

Product Code. 11430788
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430788 5 g 5g
11440788 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430788 Supplier Thermo Scientific Alfa Aesar Supplier No. B25249.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 454-90-0
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.14
MDL Number MFCD00000385
InChI Key XHONYVFDZSPELQ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl anisole, 1-methoxy-3-trifluoromethyl benzene, 3-methoxybenzotrifluoride, 3-trifluoromethylanisole, benzene, 1-methoxy-3-trifluoromethyl, 3-methoxy-1-trifluoromethyl benzene, pubchem2712, 3-trifluoromethylanisol, acmc-1cuj0, m-trifluoromethyl anisole
PubChem CID 96691
IUPAC Name 1-methoxy-3-(trifluoromethyl)benzene
SMILES COC1=CC=CC(=C1)C(F)(F)F

Spécification

Boiling Point 159°C to 160°C
Flash Point 48°C (118°F)
Quantity 5 g
UN Number UN1993
Beilstein 3048340
Formula Weight 176.14
Percent Purity 99%
Chemical Name or Material 3-(Trifluoromethyl)anisole

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.