Learn More
3-Propanoylbenzonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Propanoylbenzonitrile |
| CAS | 50916-34-2 |
| Molecular Formula | C10 H9 N O |
| Quantity | 50 mg |
| Synonym | 3-(1-Oxopropyl)benzonitrile, 1-(3-Cyanophenyl)-1-propanone, 3-Propionylbenzonitrile, 3'-Cyanopropiophenone |
| InChI Formula | InChI=1S/C10H9NO/c1-2-10(12)9-5-3-4-8(6-9)7-11/h3-6H,2H2,1H3 |
| SMILES | CCC(=O)c1cccc(c1)C#N |
| IUPAC Name | 3-propanoylbenzonitrile |
| Molecular Weight (g/mol) | 159.185 |
| Formula Weight | 159.068 |