Learn More
3-Phenoxybenzaldehyde, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 39515-51-0 |
| Molecular Formula | C13H10O2 |
| MDL Number | MFCD00003353 |
| Quantity | 10 g |
| SMILES | C1=C(C=CC=C1OC2=CC=CC=C2)C=O |
| Molecular Weight (g/mol) | 198.22 |
| Percent Purity | 95% |
| Chemical Name or Material | 3-Phenoxybenzaldehyde |