Learn More
3-Oxo-N-phenyl-4-aza-5α-androst-1-ene-17β-carboxamide, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Oxo-N-phenyl-4-aza-5Alpha-androst-1-ene-17Beta-carboxamide |
| CAS | 158522-79-3 |
| Molecular Formula | C25 H32 N2 O2 |
| Quantity | 100 mg |
| Synonym | 1H-Indeno[5,4-f]quinoline-7-carboxamide','2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-N-phenyl-','(4aR,4bS,6aS,7S,9aS,9bS,11aR)-, 4-Azaandrost-1-ene-17-carboxamide','3-oxo-N-phenyl-','(5α,17β)-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-Tetradecahydro-4a,6a-dimethyl-2-oxo-N-phenyl-1H-indeno[5,4-f]quinoline-7-carboxamide, 1H-Indeno[5,4-f]quinoline-7-carboxamide','2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-N-phenyl-','[4aR-(4aα,4bβ,6aα,7α,9aβ,9bα,11aβ)]-, 3-Oxo-N-phenyl-4-aza-5α-androst-1-ene-17β-carboxamide |
| InChI Formula | InChI=1S/C25H32N2O2/c1-24-14-12-19-17(8-11-21-25(19,2)15-13-22(28)27-21)18(24)9-10-20(24)23(29)26-16-6-4-3-5-7-16/h3-7,13,15,17-21H,8-12,14H2,1-2H3,(H,26,29)(H,27,28)/t17-,18-,19-,20+,21+,24-,25+/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CC[C@H]4NC(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2C(=O)Nc5ccccc5 |
| IUPAC Name | (1S,3aS,3bS,5aR,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-N-phenyl-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide |
| Molecular Weight (g/mol) | 392.53 |
| Formula Weight | 392.2464 |