missing translation for 'onlineSavingsMsg'
Learn More

3-O-(2-Aminoethyl)-25-hydroxyvitamin D3, MedChemExpress

Product Code. p-200101112
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16496398

Brand: Medchem Express HY15254

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3-O-(2-Aminoethyl)-25-hydroxyvitamin D3 is a Vitamin D3 derivative.

3-O-(2-Aminoethyl)-25-hydroxyvitamin D3 is a Vitamin D3 derivative.

Specifications

Chemical Name or Material 3-O-(2-Aminoethyl)-25-hydroxyvitamin D3
CAS 163018-26-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H51NO2
Quantity 10 mg
Synonym 25-Hydroxy Vitamin D3 3,3’-Aminopropyl Ether
Solubility Information DMSO : ≥ 100 mg/mL (218.47 mM)
SMILES C=C1CC[C@H](OCCCN)C/C1=C/C=C2[C@@](CC[C@]3([H])[C@@H](CCCC(C)(C)O)C)([H])[C@]3(C)CCC/2
Molecular Weight (g/mol) 457.73
Formula Weight 457.73
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.