missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenyl isothiocyanate, 97%

Product Code. 11347716
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347716 5 g 5g
11337716 1 g 1g
11357716 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11347716 Supplier Thermo Scientific Alfa Aesar Supplier No. L11335.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3529-82-6 | C7H4N2O2S | 180.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3529-82-6
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.18
MDL Number MFCD00041245
InChI Key OEZXLKSZOAWNJU-UHFFFAOYSA-N
Synonym 3-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-3-nitro, m-nitrophenyl isothiocyanate, 3-nitrophenylisothiocyanate, isothiocyanic acid, m-nitrophenyl ester, 1-isothiocyanato-3-nitro-benzene, m-nitrophenylisothiocyanat, acmc-1co3y, phenyl, 3-nitro, isothiocyanate, #
PubChem CID 19050
IUPAC Name 1-isothiocyanato-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(=C1)N=C=S

Specifications

Melting Point 56°C to 58°C
Odor Irritating
Quantity 5 g
UN Number UN2923
Beilstein 2094061
Formula Weight 180.19
Percent Purity 97%
Chemical Name or Material 3-Nitrophenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.