missing translation for 'onlineSavingsMsg'
Learn More

3-Morpholinophenol, 97%, Thermo Scientific™

Product Code. 10243622
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10243622 10 g 10g
10415532 1 g 1g
10669495 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10243622 Supplier Thermo Scientific Maybridge Supplier No. TL00276EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 27292-49-5
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00051675
InChI Key BMGSGGYIUOQZBZ-UHFFFAOYSA-N
Synonym 3-morpholinophenol, 3-morpholin-4-yl phenol, phenol, 3-4-morpholinyl, 3-4-morpholinyl phenol, 3-morpholin-4-yl-phenol, n-3-hydroxyphenyl morpholine, m-morpholinophenol, 3-morpholinylphenol, 3-morpholino phenol, 3-morpholinyl-phenol
PubChem CID 141343
SMILES OC1=CC=CC(=C1)N1CCOCC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
IUPAC Name 3-(morpholin-4-yl)phenol
Formula Weight 179.22
Percent Purity 97%
Chemical Name or Material 3-Morpholinophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.