missing translation for 'onlineSavingsMsg'
Learn More

3-Methylxanthine, TRC

Product Code. 17903475
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17903475 1 g 1g
30132655 10 g 10g
30133559 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17903475 Supplier Toronto Research Chemicals Supplier No. TRCM3385701G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1076-22-8
Molecular Formula C6 H6 N4 O2
Molecular Weight (g/mol) 166.14
Synonym Theophylline-Ethylenediamine Imp. B (EP), Pentoxifylline Imp. B (EP), Theophylline Imp. B (EP), 1H-Purine-2,6-dione, 3,7-dihydro-3-methyl- (9CI), Xanthine, 3-methyl- (6CI,7CI,8CI), 3,9-Dihydro-3-methyl-1H-purine-2,6-dione, 3-Methyl-1H-purine-2,6(3H,7H)-dione, 3-Methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione, 3-Methyl-2,3,6,9-tetrahydro-1H-purine-2,6-dione, 3-Methyl-3,7-dihydro-1H-purine-2,6-dione, 3-Methyl-3,7-dihydropurine-2,6-dione, 3-Methyl-3,9-dihydro-purine-2,6-dione, 3-Methyl-7H-purine-2,6-dione, 3-Methylxanthine, NSC 515466, 1H-Purine-2,6-dione, 3,9-dihydro-3-methyl-
IUPAC Name 3-methyl-7H-purine-2,6-dione
SMILES CN1C(=O)NC(=O)c2[nH]cnc12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H6N4O2/c1-10-4-3(7-2-8-4)5(11)9-6(10)12/h2H,1H3,(H,7,8)(H,9,11,12)
Formula Weight 166.0491
Chemical Name or Material 3-Methyl-3,7-dihydro-1H-purine-2,6-dione (3-Methylxanthine)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.