Learn More
3-Methylthiophene-2-carboxamide, 99%, Thermo Scientific™
Product Code. p-7047224
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304915 1 g 1g
11314915 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11304915 Supplier Thermo Scientific Alfa Aesar Supplier No. L18092.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 76655-99-7
Molecular Formula C6H7NOS
Molecular Weight (g/mol) 141.188
MDL Number MFCD01208055
InChI Key QZDTWJRYMXQXBX-UHFFFAOYSA-N
Synonym 3-methyl-2-thiophenecarboxamide, pubchem16135, 2-thiophenecarboxamide,3-methyl, 3-methyl-thiophene-2-carboxamide, 3-methyl-2-thiophenecarboxamide #, 2-thiophenecarboxamide, 3-methyl-9ci
PubChem CID 588128
IUPAC Name 3-methylthiophene-2-carboxamide
SMILES CC1=C(SC=C1)C(=O)N
Specifications
Melting Point 122°C to 125°C
Quantity 1 g
Beilstein 116188
Formula Weight 141.19
Percent Purity 99%
Chemical Name or Material 3-Methylthiophene-2-carboxamide